Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher BioReagents™ 2XYT Broth (Capsules)
Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.
Specifications
Form | Solid |
---|---|
Format | Capsule |
Media Type | 2XYT |
Product Type | Microbiology Media |
Formulation | Casein Peptone 16g/L, Yeast extract 10g/L, Sodium Chloride 5g/L |
Catalog Number | Form | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Form | Quantity | Price | Quantity & Availability | |||||
BP9736-500
|
Solid | 32 x 15.5 g | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
BP97365
|
Solid | 500 mL Capsules, 5 kg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Directions for Media Preparation: Add 2 capsules per 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water). Heat and frequently agitate to completely dissolve capsule. Before use, sterilize by autoclaving for 15 minutes.
- Product is in capsule form
- Each capsule contains 15.5g of broth
Specifications
Solid | |
2XYT | |
Casein Peptone 16g/L, Yeast extract 10g/L, Sodium Chloride 5g/L |
Capsule | |
Microbiology Media |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title