Learn More
Thermo Scientific™ Finntip™ Flex Filtered Pipette Tips
Minimize the risk of cross-contamination with these flex filtered pipette tips that combine the ergonomics of a standard pipette tip with the safety of a filter tip.
Supplier:  Thermo Scientific™ 94056810
 View more versions of this product
Catalog No. 14-386-208
  • $354.65 / Case of 960
    Save 25%
    Reg : $476.00
    The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Edge
Add to Cart
Description
Description
Minimize the risk of cross-contamination with Thermo Scientific™ Finntip™ Flex Filter Pipette Tips, which combine the ergonomics of a standard Finntip Flex pipette tip and the safety of a filter tip. Offered sterile, and free from DNA, DNase and RNase. Ideal for PCR and pipetting of radioactive and infectious samples. The filters, made of an inert nonsealing polyethylene matrix, are effective at eliminating carryover contamination from liquids and aerosols between pipetter and tip.
Ordering Information:
Supplied in vacuum-sealed sterilized tip racks.
Recommended for:
Use in sensitive applications such as PCR and pipetting of radioactive and infectious samples.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Finntip Flex Filter 1200µL, sterile
Filtered
8.7cm
Turquoise
FinnTip Flex
1200 μL
Sterile
Rack
Thermo Scientific Finnpipette
Sterile & PCR Ready
Product Suggestions
Product Suggestions