Learn More
Thermo Scientific™ FinnTip Filtered Pipette Tips PROMO
Protect pipettes and the samples from contamination with these filtered pipette tips.
Supplier: Thermo Scientific™ 21377387S24
Description
Features:
- Inert and nonsealing polyethylene filter
- Tips come vacuum sealed in irradiation-sterilized hinged-tip racks
- Not autoclavable
- Certified to be free from DNA, DNase, RNase and endotoxin
Specifications
50 μL | |
Sterile | |
4.8 cm | |
10 x 384 Tips/Rack | |
Single-use | |
FinnTip |
Filtered | |
Not Autoclavable | |
Rack | |
Turquoise | |
Thermo Scientific Finnpipette | |
Sterile & PCR Ready |