Learn More
Tritech Forensics Inc Fingernail Clippings Evidence Collection Kit
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Tritech Forensics Inc FINCL0GEN
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
During an assault, the victim's fingernail(s) may break off during the struggle. Our Fingernail Clippings Evidence Collection Kit is used to collect the remaining nail for comparison with the broken off fingernail found at the scene of the assault to confirm the victim's description of where the assault took place. When the clipping(s) is submitted to the crime laboratory, trace amounts of the assailant's DNA may be found on the clipping and on the recovered broken nail as well.Kit Components:1 ea. Tamper-evident, pre-sealed Kit Envelope, 5.5" x 7.5". 1 ea. Kit Instruction Sheet. 1 ea. Metal Fingernail Clipper. 1 ea. Right-Hand Envelope containing one paper bindle. 1 ea. Left-Hand Envelope containing one paper bindle. 1 ea. Tamper-evident Police Evidence Seal for resealing kit envelope after specimen collection. Sold 25/case
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.