Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Excelta™ Fine Blade Straight Stainless Steel Scissors
Designed for the fine detail dissection work.
Supplier: Excelta™ 275
Description
- Straight
- Stainless Steel
- Acid resistant
- Rustproof
- Autoclavable
Specifications
Straight Scissors | |
4.5 in. | |
28 mm | |
Autoclavable | |
1.10 in. |
Very Fine | |
113 mm | |
Stainless Steel | |
Delicate applications |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction