Learn More
Thomson Instrument Company 5 Position Filter Vial Toggle Press | EA
Supplier: Thomson Instrument Company 35005
The Thomson Filter Vial Press enables high solid content and viscous liquids to be easily filtered through vials. Some fermentation cultures that reach 100 OD or particulate laden samples may require the toggle press. Filter vial presses are available in 5 and 48-vial configurations. The press allows for consistent ergonomic use of filter vials. The 48-position block can be transitioned to some automation and autosampler systems directly. Thomson manufactures its products in the USA to assure same day shipping.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.