Learn More
Basix™ Filter Tips
The Basix™ pipette tip portfolio consists of the most commonly used tip volumes and packaging options in todays laboratory.
 View more versions of this product
Catalog No. 13-611-125
  • $95.65 / Pack of 1000
    Save 8%
    Reg : $104.00
    The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $472.65 / Case of 5 PK
    Save 8%
    Reg : $514.00
    The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Description
Description
The Basix™ pipette tip portfolio consists of the most commonly used tip volumes and packaging options in todays laboratory.
Basix offers reliability and quality for all varieties of experiments, from everday liquid transfer to PCR and Electrophoresis, Basix offers a tip to meet your needs.
Choose from non-Sterile or Sterile with packaging options including bulk, racked, reloads, stack racks.
  • Unique small micron filter prevents aerosol contamination with no obstruction to airflow
  • Use aerosol barrier tips to ensure result integrity
  • Certified by independent laboratory sterile;
  • RNase, DNase, DNA, and pyrogen free
  • Natural color
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1000 μL
Sterile
Not Autoclavable
7.93cm
Rack
Universal Fit Pipettor
Filtered
Natural
3.125 in.
DNase, RNase, DNA, and Pyrogen Free
Universal
Basix
Product Suggestions
Product Suggestions