Learn More
Microsolv Technology Corporation Filter Sf .45 Regcel 4Mm 100Pk
Supplier:  Microsolv Technology Corporation 58045R04C
Encompass_Preferred
Syringe Filters, 4mm, RC, 0.45um Pore Size. Brown Polypropylene, AQ Brand. 100/PK.
Catalog No. 50-014-52614