Learn More
Andwin Scientific FILTER SAMPLE FOR COBAS 1000
Supplier: Andwin Scientific 2083049030
The Surety� column II is compatible to use with sample cups directly or with carriers. This product has a volume capacity of 5 mL. The column is ideal for eliminating fibrin and macro particles from serum in order to remove the risk of line clogging. This is sold in a case of 1000.