Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Filter Paper, Rectangular, Grade 1
SDPSCI_ED

Economical filter paper for filtering applications

$46.38 - $66.44

Specifications

Format Rectangle
For Use With (Equipment) Conical Funnel
Material Paper
View More Specs

Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
S15674
SDPSCI_ED
View Documents
United Scientific™
FPR8X11
8.5 x 11 in.
Pack of 100 for $46.38
Add to Cart
 
S15846
SDPSCI_ED
View Documents
United Scientific™
FPR18X22
18 x 22 in.
Pack of 100 for $66.44
Add to Cart
 
Description

Description

  • Pack of 100 filter papers.
  • Available in 18 x 22 in. and 8.5 x 11 in.
  • Grade 1 filter papers feature medium porosity and flow rate and are the most used for routine applications.
Specifications

Specifications

Rectangle
Conical Funnel
Paper
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit