Learn More
United Scientific™ Filter Paper, Circular
Economical filter paper for filtering applications
Supplier: United Scientific™ FPR240
Description
- Grade 1 filter papers feature medium porosity and flow rate and are the most commonly used for routine applications.
- Pack of 100 circular filter papers
- Offered sizes: 7, 9, 11, 12.5, 15, 18, 24, and 32 cm in diameter
Specifications
Disc | |
Conical Funnel | |
Paper | |
32 cm |