Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific™ Filter Paper, Circular
Economical filter paper for filtering applications
Supplier: United Scientific™ FPR320
Description
- Grade 1 filter papers feature medium porosity and flow rate and are the most commonly used for routine applications.
- Pack of 100 circular filter papers
- Offered sizes: 7, 9, 11, 12.5, 15, 18, 24, and 32 cm in diameter
Specifications
Disc | |
Conical Funnel | |
Paper | |
24 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction