Learn More
Revvity Health Sciences Inc Filter Count, 1 Liter
Supplier: Revvity Health Sciences Inc 6013141
Filter-Count is a pseudocumene-based LSC cocktail specifically formulated to dissolve cellulose nitrate membrane filters. Filter-Count can also dissolve mixed cellulose esters and PVC (polyvinyl chloride) filters, however these sample types may require additional time. Filter-Count will dissolve these wet or dry filters, reducing sample preparation procedures and improving counting results by enabling the use of external standard quench monitoring. Features Dissolves cellulose nitrate, mixed cellulose esters and polyvinyl chloride filters Eliminates self-absorption by the filter Enables automatic DPM determinations Accepts wet or dry filters Flash-point of approximately 50'C High counting efficiency
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.