Learn More
Parker Hannifan FILTER CARTRIDGE,0.01MIC 99.9%
Supplier:  Parker Hannifan 10018BX
Encompass_Preferred
Parker Balston Replacement Filter Cartridges, Box of 10Parker Balston Model 100-18, Filter Compressed Air and Gas, Box of 10 Tubes Replacement Elements, 99.99% at 0.01 micron, Filter Element, Grade-BX.
Catalog No. NC2226036