Learn More
Parker Hannifan FILTER CARTRIDGE,0.01 MIC 93%
Supplier:  Parker Hannifan 10018DX
Encompass_Preferred
Parker Balston Replacement Filter Cartridges, 0.01 mic 93%, Box of 10Parker Balston Model 100-18, Filter Compressed Air and Gas, Box of 10 Tubes Replacement Elements, 137 scfm at 100 psig, 93% at 0.01 micron, Filter Element, Grade-DX.
Catalog No. NC2226042