Learn More
Restek™ FID Maintenance Kits
SDP
For use with Agilent GCs
$1229.00
Specifications
Type FID Maintenance Kit
Quantity 1 Kit
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
06-611-204
SDP
View Documents
Restek™
22180
Agilent 5890 GCs
Each for $1,229.00
Add to Cart
06-611-203
SDP
View Documents
Restek™
22179
Agilent 6850/6890/7890 GCs
Each for $1,229.00
Add to Cart
Description
Description
  • Includes a variety of the most common consumable GC supplies and tools
  • All parts meet or exceed performance of instrument manufacturer's parts
  • Parts list makes reordering easy
Specifications
Specifications
FID Maintenance Kit
1 Kit