Learn More
Perkin Elmer US LLC FID Gas Station - NM Plus, 650 mL/min H2
Supplier:  Perkin Elmer US LLC N9300022
Encompass_Preferred
Request Bulk Order
Designed with space saving in mind, it sits directly under the GC to conserve bench space. You can also apply the FID gas station to GCMS systems with the simple addition of a drawer tray for the MS to rest on. If you want to benefit from the space saving design, but only require a H2 generator, that is also an option. H2 Flow Rate: 650 mL/min Zero Air Flow Rate: 5 L/min Zero Air Purity: <0.1 ppm Certifications: CE, CSA, FCC
Catalog No. 50-210-5458