Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Fibre-Metal™ Welder's Cover Plates
Click to view available options
2W x 4.25 in. L
Description
Fibre-Metal provides safety and cover plates for welding helmets and goggles. The HIGH PERFORMANCE™ line includes a top quality cover plate for virtually every metal fabricating application. All products are produced to the highest standards with rigid quality control to enhance a fabricator's view of the job for maximum productivity while providing protection in accordance with ANSIZ87.1 requirements.
Specifications
Lens Tint | Clear |
Product Type | Cover Plate |
Length (English) | 4.25 in. |
Width (English) | 2 in. |