Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Fibre-Metal™ Face Shield Headgear
Click to view available options
Includes:
Crown with Mounting Cups
Faceshield Adaptor
Headgear Only
Description
- Window is not included and sold separately
- Quick-Lok™ Cups can be can be securely mounted onto, and removed from the cap without tools
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Width (English) | 3 in. |
Certifications/Compliance | ANSI Z87.1-2003 |
Color | Gray |
Product Type | Faceshield Headgear |
For Use With (Equipment) | Fits Over Large Sized Spectacles |
Includes | Headgear Only |
Material | HDPE, Noryl™ |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction