Learn More
Dolan-Jenner Industries FIBER-LITE MI-LED W/DUAL FIBER
Supplier: Dolan-Jenner Industries MILEDUSDG
Dolan-Jenner Fiber-Lite LED illuminator with dual gooseneck fibers and lenses, 115v cord, 40,000 hour life, higher output than 150w halogen sources, CE, ETL, RoHS, Made in USA