Learn More
Thermo Scientific CE Phosphoramidite, dC(Bz), 1 g
Supplier: Thermo Scientific 400328
Chemical Name: dC(Bz) CEP. The dC(Bz) CE Phosphoramidite is used for nucleic acid synthesis on ABI DNA Synthesizers. Standard series of phosphoramidite deprotects in 6-8 hours at 55°C.