Learn More
QA Bio Inc Fetuin N Glycan Library
Supplier:  QA Bio Inc CLIBNFETUIN01
Encompass
The fetuin N-linked glycan library in this product have been released from 50 µgs of a fetuin standard that is purified from fetal calf serum resulting in mixture of bi-, tri-, and tetra-antennary glycan standards with variable sialylation. A fetuin O-linked glycan library (CLIBO-FETUIN-01) and a fetuin glycoprotein (GCP-FET-05) are also available.
Catalog No. 50-223-6572