Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Fetal Bovine Serum, qualified, heat inactivated, United States
Important Update: We've recently introduced Premium Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Qualified US origin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.
Supplier: Gibco™ A3840102
Description
- Offers excellent value for basic cell culture, specialty research, and specific assays
- It has earned the trust of researchers with consistent quality and award-winning support that helps meet research needs and budget requirements
- Heat-inactivated serum eliminates the multiple steps necessary to heat inactivate
- It improves product integrity and consistency since the heat inactivation is done in a controlled environment
- ISO 13485-certified, processed in FDA-registered facilities
- Labeled for in vitro diagnostics (compliant with highest level of USP sterility testing)
Additional Information
- Serum category: Value Plus (Performance)
- Serum for general cell culture with common cell lines
- Triple filtered at 0.1 μm
- Endotoxin level: ≤10 EU/mL
- Hemoglobin level: ≤ 25 mg/dL
- Origin: United States
One-Shot Format Benefits
- Wide mouth, chamfered edge, and curved shape for easy pouring and pipetting
- Engineered to freeze/thaw without splitting or breaking
- Fits in test tube racks and frees up freezer space
- No labeling mishaps with our easy-to-read package label containing all of the information you need
- 33% less waste generated than aliquoting from 500-mL bottles
Order Info
Shipping Condition: Dry Ice
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
Bovine | |
One Shot | |
10 x 50 mL | |
Sterile | |
≤ 10 EU/mL | |
Qualified | |
Frozen |
Fetal Bovine Serum, heat inactivated, qualified, One Shot™, United States | |
United States | |
Fetal Bovine Serum | |
Heat-inactivated | |
Triple-filtered, 0.1 μm | |
≤ 25 mg/dL | |
One Shot™ |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction