Learn More
Gibco™ Fetal Bovine Serum, Premium Plus, heat inactivated
Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.
Specifications
Content And Storage | Store at ≤-10°C. |
---|---|
Age | Fetal |
Species | Cattle/Bovine |
Country of Origin | Refer to CoA for specific origin |
Form | Frozen |
Catalog Number | Mfr. No. | Quantity | Format | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Format | Price | Quantity & Availability | ||||
A5669501
|
Gibco™
A5669501 |
50 mL | One Shot |
Each for $114.00
|
N/A | ||||
A5669502
|
Gibco™
A5669502 |
10 x 50 mL | One Shot |
Pack of 10 for $1,072.00
|
N/A | ||||
A5669801
|
Gibco™
A5669801 |
500 mL | Bottle |
Each for $908.00
|
N/A | ||||
Description
Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.
Sera category: Premium Plus
- Use for a broad range of cell types, especially sensitive cell lines
- Our most characterized serum with the lowest endotoxin and hemoglobin levels
- Endotoxin level: ≤5 EU/mL
- Hemoglobin level: ≤20 mg/dL
- Origin: Refer to CoA for specific origin (US, Australia, New Zealand)
- US Department of Agriculture (USDA) safety tested for the absence of bluetongue and akabane from countries that are USDA-approved for importation into the US
Gibco Serum Delivers
- ISO 13485-certified; processed in FDA-registered facilities
- Triple filtered at 0.1 μm
- Gibco bottle easier to use in hood, reduces risk of contamination and helps improve consistency
- Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook
Specifications
Store at ≤-10°C. | |
Cattle/Bovine | |
Frozen | |
Standard (Sterile-filtered) | |
Triple-filtered, 0.1 μm | |
Triple-filtered, 0.1 μm | |
≤5 EU/mL | |
Yes | |
Dry Ice |
Fetal | |
Refer to CoA for specific origin | |
Fetal Bovine Serum | |
Sterile | |
Premium Plus | |
Up to 70 quality tests including EMA virus testing; USP/EP mycoplasma, endotoxin, performance; biochemical/hormonal profiling; Oritain fingerprinting | |
Less waste | |
≤20 mg/dL | |
USDA Safety Testing Performed for Australian origins |
Safety and Handling
ShelfLife : 5 Years
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.