Learn More
World Precision Instrument Femtojet MICRO-ePORE Holder, Platinum Wire
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: World Precision Instrument 300684PT
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
These electrode holders are designed exclusively for use the with MICRO-ePORE™ system, which includes the silver wire electrode holders. The platinum wire electrode holders are an optional accessory. Platinum wire electrode holders are important when you are working with proteins. The silver chloride wire is perfect for work with RNA. However, the silver chloride wire tends to denature proteins, reducing the efficiency of the microinjection solution.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.