Learn More
Cole-Parmer Cole-Parmer BNC-to-Stripped Wire Adapter
Supplier: Cole-Parmer 5610560
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Use this female BNC-to-stripped wire end connector to simplify connection and replacement of pH/ORP electrodes with male BNC connections to transmitters and controllers with terminal block connectors.