Learn More
Med Vet International Felocell 4, Feline Vaccine, Modified Live Virus, 25 x 1mL
Supplier:  Med Vet International PHVFELOCELL4
Encompass_Preferred
Feline Rhinotracheitis-Calici-Panleukopenia-Chlamydia Psittaci Vaccine, Modified Live Virus and Chlamydia, cats. Preservative: Gentamicin
Catalog No. 50-218-8540