Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3B Scientific™ Feline Elbow-Shoulder Model
3B Scientific Feline Elbow-Shoulder Model features normal bone and osteoarthritic changes to joints.
Supplier: 3B Scientific™ 1019588
This item is not returnable.
View return policy
Description
- Average size feline elbow and shoulder features: normal bone and osteoarthritic changes to joints.
- Model includes the following bones: scapula, humerus, radius, ulna and nine important ligaments and tendons.
- Model size: 7.75 x 4 x 7/8 in.
- Base: 6.5 x 5 in.
Specifications
Feline Model | |
6.5 x 5 in. Base | |
Scapula, Humerus, Radius, Ulna and Nine Ligaments and Tendons |
7.75 x 4 x 7/8 in. | |
Anatomy Demonstrations | |
0.44 lb. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction