Learn More
Med Vet International 8Fr Feeding Tube, Non-Sterile, Each
Supplier: Med Vet International FT8
Red Rubber Latex Feeding Tube. Smooth tip, two oval eyes and funnel end. 16" length and radiopaque.