Learn More
Medex Supply FEATHER ST DISP SCAPELS 11 20P
Supplier: Medex Supply GRF2975#11
eather Sterile Scalpels, Disposable #11, 20/bx. Feather Model: 2975#11. Medex SKU: GRF-2975#11. With one of the largest catalogs of medical, surgical and diagnostic supplies available online, Medex Supply can accommodate your facility's needs for Surgical Supplies. Scalpels, Blades & Knives can be found in our extensive online collection of products from globally recognized and trusted brands, including Feather. an excellent option to consider is the Sterile Scalpels. Sterile, disposable scalpel features heat-treated stainless steel blade securely integrated with rigid, chemically resistant plastic handle. Individually wrapped in hermetically-sealed aluminum foil. Blades are protected with snap-off cover.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.