Learn More
Medex Supply FEATHER DISP. SCAPELS NO10
Supplier: Medex Supply GRF2975#10
Feather Scalpels #10 are sterile, disposable scalpel features heat-treated stainless steel blade securely integrated with rigid, chemically resistant plastic handle. Individually wrapped in hermetically-sealed aluminum foil. Blades are protected with snap-off cover. Heat-treated stainless steel blade securely integrated with rigid, chemically resistant plastic handle. 20/PK.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.