Learn More
Perkin Elmer US LLC Iron (Fe) Lumina Hollow Cathode Lamp
Supplier: Perkin Elmer US LLC N3050126
Single Element Lumina hollow cathode lamp (HCL) for the detection of elemental Iron (Fe). Every genuine PerkinElmer lamp is designed for use with and tested on PerkinElmer spectrometers to assure compatibility and the highest performance. PerkinElmer offers a wide range of single-element and multi-element HCLs, which are ideal for determining most elements by atomic absorption spectroscopy. To prolong the life of a Lumina Hollow Cathode Lamp (HCL), we produce lamps with larger internal volume so that a greater supply of fill gas at optimum pressure is available. The larger the lamp, the greater the inert gas volume — and the longer the lamp lifetime. Note: Lumina hollow cathode lamps may also be used with all earlier AA spectrometers that do not have automatic code reading capability by using an appropriate adapter cable.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.