Learn More
Cytiva FDII DFL MEMBR-KP-1/2IN HB CON
Supplier:  Cytiva KA2DFLP6S
Encompass
: Fluorodyne II DFL membrane in Kleenpak capsule, 0.2 m sterilizing grade, 800 cm EFA, in. hose barb connections, pre-sterilized by gamma irradiation
Catalog No. NC2173641