Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Fat Diet, High, Modified
Fat Diet, High, Modified is a diet for studying obesity in animal models.
$4872.00 - $11870.00
Catalog Number | Mfr. No. | Packaging Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Packaging Quantity | Price | Quantity & Availability | |||||
02-960-19220
|
MP Biomedicals™
0296019220 |
20 kg |
Each for $4,872.00
|
|
|||||
02-960-19250
|
MP Biomedicals™
0296019250 |
50 kg |
Each for $11,870.00
|
|
|||||
Description
- Normal High Fat Diet modified to contain essential trace minerals, DL-Methionine and Fiber.
- Fat content is 45% fat (product must be stored before use).
- Typically provides 5.81 kcal/gm diet.
Specifications
Fat Diet | |
Pellets |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title