Learn More
Research Products International Corp Fastwells, 9mm Dia, 10ul, 50/pk
SDP
Supplier:  Research Products International Corp 248935
Encompass_Preferred
Fastwells form hydrophobic reagent barriers around various size specimens. Ideal for rapid isolation of cells and tissues on slides or coverslips for antibody incubations. Fastwells remain sealed during agitated washing steps to prevent leakage. High tacky silicone material peels off quickly without leaving a residue. Fastwells can be washed and autoclaved.
Catalog No. 50-197-7674