Learn More
Research Products International Corp Fast-Release Pipette Pump, Blue, 2ml
Supplier: Research Products International Corp 437902
Pipette Pump provides easy and accurate one-hand pipetting. Specially designed to offer precise alignment, secure grip and locking of glass or plastic pipettes. Features rapid release lever for quick emptying and thumbwheel for drawing and dispensing liquid.