Learn More
Roche Diagnostics KAPA SYBR FAST One-Step qRT-PCR Kits - 100 x 20 uL reactions, 1 mL
Supplier: Roche Diagnostics 07959613001
KAPA SYBR FAST One-Step qRT-PCR Kits are designed for optimal performance in stringent qRT-PCR reaction conditions, exhibiting improvements in sensitivity, specificity, reaction efficiency, and fluorescence. The kits contain an M-MuLV reverse transcriptase, an RNase inhibitor, and a novel DNA polymerase engineered through our directed evolution technology. *This item is non returnable.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.