Learn More
Falcon Safety Dust-Off™ Classic Duster
Removes dust and lint from computers, electronics, cameras, keyboards, workshops and more
$292.00 - $312.00
Specifications
For Use With (Application) | Removes dust and lint from computers, electronics, cameras, keyboards, workshops |
---|---|
Volume (English) | 10 oz. |
Volume (Metric) | 0.29 L |
Alert:
Keep away from heat, open flames, or other sources of ignition.
Description
- Chrome valve for dependable use
- 100% ozone safe
- Contains bitterant to discourage inhalant abuse
- 10 oz. can
Specifications
Removes dust and lint from computers, electronics, cameras, keyboards, workshops | |
10 oz. | |
0.29 L |
Use good industrial hygiene practices in handling this material. Avoid contact with eyes and skin. Avoid breathing mists or aerosols of this product. Use only with adequate ventilation. Flammable gas may catch fire. Contact with liquid may cause frostbite. Excessive intentional inhalation may cause respiratory tract irritation and central nervous system effects (headache, dizziness). Vapors may cause dizziness or suffocation.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.