Learn More
Glas Col LLC Heating Mantle
Supplier: Glas Col LLC 100A O963
Fabric mantle for 100ml spherical flask, 80W lower, top not heated, completely covers top and bottom of flask. To 400 degrees C internal operating temperature. Top and bottom held together with zipper to prevent heat loss. The side split in the top allows removal without disturbing the flask.