Learn More
MilliporeSigma™ Supelco™ EZSafe™ Flash-Chromatography Column with Reservoir and Fritted Disc
Flash chromatography is useful for rapid, preparative separations with moderate resolutions.
$584.00
Specifications
Column Type | Flash Chromatography |
---|---|
For Use With (Application) | Flash Chromatography |
Product Line | EZSafe |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
11-100-8888
|
MilliporeSigma™ Supelco™
Z4159871EA |
600 mL |
Each of 1 for $584.00
|
|
|||||
Description
Low pressure (max. 7 psig) medium wall columns with reservoir. Greaseless PTFE insert with GL-45 screw-thread attaches flow control valve to column. Precision all-PTFE valve and drip-tip.Specifications
Flash Chromatography | |
Flash Chromatography | |
EZSafe |
EZ Safe is a registered trademark of Gerresheimer Glass, Inc.