Learn More
Air-Tite EZ FLO Hypodermic Needle
Smooth entry, smooth delivery
Supplier: Air-Tite NEZ22300
This item is not returnable.
View return policy
Description
- Air-Tite Product's newest addition, the EZ FLO line of hypodermic needles, offer high quality and a broad selection at an affordable price.
- Ranging from 18 gauge to 30 gauge, these needles offer a standard bevel, thin wall from 18 g to 25 g and regular-wall in 27 g and 30 g to combine comfort and durability.
- Each needle is sterile, individually packaged in a box of 100.
- Made of surgical stainless steel and a polypropylene hub.
Specifications
Surgical Stainless Steel | |
22 G | |
100/Box |
Gray | |
3 in. | |
Sterile |