Learn More
GUARDIAN EQUIPMENT Bacteriostatic Additive (Case of 4)
Supplier: GUARDIAN EQUIPMENT G1540BAR
8 oz. bottle of bacteriostatic additive to protect against growth of bacteria, fungus, algae, and acanthamoeba in portable eyewash units. (pkg of 4)