Learn More
American Educational Products Eye Model Activity Set
Let's go on a trip inside the eye!
Supplier: American Educational Products 2640
Includes: Model and comprehensive activity guide.
Description
This large, three-piece model of the human eye can be taken apart to show the body of the eye, the lens, and a multicolored cover depicting veins, sclera and cornea. A separate diagram panel shows the parts of the retina. Dimensions: 18H × 24 in.W.