Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Phoenix™ Extreme Weather Components

Replacement parts for Educational Kit

Specifications

Grade K to 5
For Use With (Application) Education Kit
View More Specs

Description

Description

Variety of components specially formatted as replacements for AquaPhoenix custom designed hands-on educational kits.
Specifications

Specifications

K to 5
Education Kit
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit