Learn More
Research Products International Corp eXTReme Seal, Large, Clear, Non-sterile, 100/pk
Supplier: Research Products International Corp 202561
eXTReme is ideal for PCR, light sensitive samples, robotics, coldroom and longterm storage. Offers universal fit for use with all plate types, minimizes sample evaporation and reduces well-to-well cross contamination. Specially formulated adhesive provides excellent seal for all plates. Removes cleanly from plate with no adhesive residue and does not tear upon removal. Smaller dimensions and chamfered corners result in no plate overhang for robotics. eXTReme is a Trademark of Excel Scientific, Inc.
PCR and longterm storage
RNase, DNase free
Withstands -40° to +120°C
Pierceable, autoclavable
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.