Learn More
Research Products International Corp eXTReme Foilseal, Large, Non-S, 100/pk
SDP
Supplier:  Research Products International Corp 202549
Encompass_Preferred
eXTReme Seal is ideal for  light sensitive samples, robotics, coldroom and longterm STOR. Offers universal fit for use with all plate types, minimizes sample evaporation and reduces well-to-well cross contamination. Specially formulated adhesive provides excellent seal for all plates. Removes cleanly from plate with no adhesive residue and does not tear upon removal. Smaller dimensions and chamfered corners result in no plate overhang for robotics. eXTReme is a Trademark of Excel Scientific, Inc.
Catalog No. 50-223-0155