Learn More
Air Impurities Removal Systems Extract-ALL™ Extractor Arms
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Air Impurities Removal Systems E9823
Includes: Includes hose clamp, rubber clamp cover, 24 in. (61cm) long wall-mounting swivel boom
Description
Efficient way to capture chemical dust fumes, soldering fumes, solvent odors, dust from sanding and mixing, and a variety of other particulates.
- Self-supporting, articulating arms constructed from durable polypropylene
- Resist abrasion, heat and chemicals
- Complete extractor arm system with a 120V Extract-ALL Exhaust Blower
Order Info
2.5 in. (6.4cm) diameter; expandable range of 36 to 53 in. (91 to 135cm)
Certifications
UL listed, UL listed and CSA approved.
Specifications
Polypropylene |