Learn More
Perkin Elmer US LLC External Igniter FID Catalytic Reactor Accessory
Supplier: Perkin Elmer US LLC N6120070
The catalytic reactor consists of a special catalytic reactor base which replaces the FID base. The reactor body contains a quantity of catalyst held in place by a quartz wool plug.