Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Extendable Pole XSL008, Sold by AquaPhoenix Scientific

Extendable aluminum pole with 3M™ sponge-stick holder
Supplier: 3M™ XSL008
Description
- Telescoping 8 ft 5 in. (2.56 m) pole
- Includes 3M™ sponge-stick holder
Specifications
Extendable Sponge Pole | |
2.56 m | |
Sponge stick holder |
8.42 ft. | |
Aluminum |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title