Learn More
Cole-Parmer Extech VIR50 Video Infrared (IR) Thermometer
Supplier: Cole-Parmer 3975308
The unique combination of a infrared thermometer and digital camera allows the user to actually see what the thermometer is measuring. The large 2.2" color video display, shows both temperature and humidity as it relates to a specific point on a displayed photo. Target data along with either still photos or videos can be recorded onto a standard SD memory card, instead of a built-in memory. The advantage of the data being stored on an SD card is when the memory is full the SD card can be simply exchange out for unlimited data storage. The removable SD card is easy to transport and insert into a computer’s memory card slot or SD card reader for upload. The data is provided in Excel, allowing the user to easily analysis the information without the need for additional software. Other features include min/max, hold, temperature differential, offset adjustment, adjustable emissivity, type K input, alarm, date and time stamp, auto power off and low battery indicator.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.